missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Death Ligand Signal Enhancer Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81912PCP
This item is not returnable.
View return policy
Description
Death Ligand Signal Enhancer Polyclonal antibody specifically detects Death Ligand Signal Enhancer in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Death Ligand Signal Enhancer | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| DAP3 Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, DAP3-Binding Cell Death Enhancer 1, Long Form, Death Ligand Signal Enhancer, DELE, DELE1, DELE1(L), hypothetical protein LOC9812, KIAA0141 | |
| Death Ligand Signal Enhancer antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human Death Ligand Signal Enhancer. The immunogen is located within the first 50 amino acids of Death Ligand Signal Enhancer. Amino Acid Squence: RVTPKSTSPDGPQTTSST | |
| 0.1 mL | |
| Apoptosis | |
| 9812 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction