Description
Corning universal fit tips in hinged DeckWorks racks are available in 10 μL, 200 μL, 300 μL, and 1,000 μL sizes. The 10 μL, 300 μL, and 1,000 μL tips are extended length and allow for improved access into microcentrifuge (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes. Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 μL DeckWorks pipet tips are designed for all popular 300 μL multi-channel instruments.
Specifications
Specifications
| Volume (Metric) | 100 to 1000 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Nonpyrogenic, Human DNA-free, RNase/DNase-free, Sterility Assurance Level 10-3 |
| Clean Claims | Nonpyrogenic, Human DNA-free, RNase/DNase-free, Sterility Assurance Level 10-3, endotoxin free |
| For Use With (Application) | Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking |
| Format | Hinged Rack |
| Tip Style | Round |
| Show More |