missing translation for 'onlineSavingsMsg'
Learn More

Borer Chemie™ Deconex 12 PA-x

Product Code. 10759418
Change view
Click to view available options
Volume (Metric):
1.3 kg
Unit Size:
1300g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10759418 1.3 kg 1300g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10759418 Supplier Borer Chemie™ Supplier No. 516800.00F10W

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Deconex 12 PA-x
pH 11.7
Appearance Light Yellowish
Composition Alkalis, complexing agents, dispersing agent, Solubilizer, surface active compounds, wetting agent
For Use With (Application) Deconex 12 PA-x is a liquid, mildly alkaline yet highly efficient concentrate for the spontaneous cleaning and decontamination of delicate laboratory equipment and as well of optical and electronic components (e.g. semiconductors) out of glass, quartz, porcelain, Metal etc
Volume (Metric) 1.3 kg
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.