Learn More
Borer Chemie™ deconex 21 SOLID
Product Code. 10258370
Click to view available options
Volume (Metric):
1 x 10 kg
Unit Size:
10kg
Product Code. 10258370
Brand: Borer Chemie™ 501100.00S10P
Powder cleaner for laboratory glassware and laboratory utensils. Thanks to its excellent material compatibility, deconex 21 SOLID also cleans sensitive objects made of aluminium. An acid rinsing with a deconex cleaner is recommended.
  • Powder cleaner for automated cleaning
  • High yield, typical concentration: 3-5 g/L
  • Excellent material compatibility, also for aluminium
  • Rinses off without leaving residues
  • For consistent and flawless results
  • Wide cleaning spectrum
  • Without chlorine, EDTA, NTA and surfactants
  • Acid rinse with deconex cleaner recommended
  • Technical support by the manufacturer
Specifications
Form Powder
pH ∼12.0 (1% Solution)
Composition Alkalis, complexing agents, corrosion inhibitor
Concentration Concentrate
Container Type Bucket
Content And Storage 5°C to 25°C
Quantity 1 x 10 kg
Volume (Metric) 1 x 10 kg
Weight (English) 22 lb.
Weight (Metric) 10 kg
compliance-icons compliance-icons
Product Identifier
  • deconex 21
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : TETRAPOTASSIUM PYROPHOSPHATE, DIPOTASSIUM TRIOXOSILICATE, SODIUM HYPOCHLORITE