missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borer Chemie™ deconex 24 LIQ
Click to view available options
Quantity:
1 x 7 kg
1 x 14 kg
1 x 35 kg
Unit Size:
14kg
35kg
7kg
Description
- Concentrate for automated cleaning
- High yield concentrate, with active chlorine
- Typical application concentration: 3-5 g/L
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Wide cleaning spectrum
- Without EDTA, NTA and surfactants
- Acid rinse with deconex cleaner recommended
- Technical support by the manufacturer
Specifications
Specifications
| Form | Liquid |
| pH | ∼12.5 (1% Solution) |
| Composition | Alkalis, complexing agents, corrosion inhibitor, sequestering agent, inorganic chlorine donor |
| Concentration | Concentrate |
| Container Type | Canister |
| Content And Storage | 5°C to 25°C |
| Quantity | 1 x 14 kg |
| Volume (English) | 2.5 gal. |
| Volume (Metric) | 9.3 L |
| Weight (English) | 30.9 lb. |
| Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction