missing translation for 'onlineSavingsMsg'
Learn More

Decoquinate, MedChemExpress

Product Code. 16428576
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16428576

Brand: Medchem Express HYB1036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Decoquinate is a quinolone derivative that can be used for research of coccidiosis in domestic ruminants. Decoquinate also has potent activity against both Plasmodium hepatic development and red cell replication.

Decoquinate is a quinolone derivative that can be used for research of coccidiosis in domestic ruminants. Decoquinate also has potent activity against both Plasmodium hepatic development and red cell replication.

Specifications

Chemical Name or Material Decoquinate
Physical Form Solid
CAS 18507-89-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C24H35NO5
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 1 mg/mL (2.39 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (2.39 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C2=CC(OCCCCCCCCCC)=C(OCC)C=C2N=C1)OCC
Molecular Weight (g/mol) 417.54
Formula Weight 417.54
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.