missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin Antibody (DCN/6289), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21077G
This item is not returnable.
View return policy
Description
Decorin Monoclonal antibody specifically detects Decorin in Human, Mouse, Porcine, Bovine, Canine, Chicken, Equine, Elk, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Decorin | |
| Monoclonal | |
| DyLight 488 | |
| Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
| Recombinant full-length Decorin protein | |
| 0.1 mL | |
| Cancer | |
| 1634 | |
| Store at 4°C in the dark. | |
| IgG1 λ |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DCN/6289 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction