missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin Antibody (DCN/8760R), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20748AF350
This item is not returnable.
View return policy
Description
Decorin Monoclonal antibody specifically detects Decorin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Decorin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
| Recombinant human Decorin protein fragment (around aa212-336) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 1634 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DCN/8760R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction