missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DECR1 Monoclonal Antibody (OTI10A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809014S
This item is not returnable.
View return policy
Description
DECR1 Monoclonal antibody specifically detects DECR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| DECR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP98 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 58529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10A11 | |
| Unconjugated | |
| MYOZ1 | |
| 1200012F07Rik; 2,4-dienoyl CoA reductase 1, mitochondrial; 2,4-dienoyl-CoA reductase [NADPH]; 2,4-dienoyl-CoA reductase 1, mitochondrial; 2,4-dienoyl-CoA reductase, mitochondrial; 4-enoyl-CoA reductase; 4-enoyl-CoA reductase [NADPH]; Decr; DECR1; Nadph; SDR18C1; Short chain dehydrogenase/reductase family 18C member 1; short chain dehydrogenase/reductase family 18C, member 1 | |
| Human recombinant protein fragment corresponding to amino acids 106-299 of human MYOZ1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction