missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DECR1 Monoclonal Antibody (OTI5D12), TrueMAB™, OriGene
Description
DECR1 Monoclonal antibody specifically detects DECR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
| Antigen | DECR1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5D12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | DECR1 |
| Gene Accession No. | Q16698 |
| Gene Alias | 1200012F07Rik; 2,4-dienoyl CoA reductase 1, mitochondrial; 2,4-dienoyl-CoA reductase [NADPH]; 2,4-dienoyl-CoA reductase 1, mitochondrial; 2,4-dienoyl-CoA reductase, mitochondrial; 4-enoyl-CoA reductase; 4-enoyl-CoA reductase [NADPH]; Decr; DECR1; Nadph; SDR18C1; Short chain dehydrogenase/reductase family 18C member 1; short chain dehydrogenase/reductase family 18C, member 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?