missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dectin-2/CLEC6A Antibody (3D1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27159JF646
This item is not returnable.
View return policy
Description
Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Dectin-2/CLEC6A | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CLEC6A | |
| A portion of amino acids 75-139 of human Dectin-2 was used as the immunogen for the antibody. 90-106 | |
| 0.1 mL | |
| 93978.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 3D1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
| CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction