missing translation for 'onlineSavingsMsg'
Learn More

Decyl Hexyl Phthalate, TRC

Product Code. 30135180
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135180 500 mg 500mg
17994055 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135180 Supplier Toronto Research Chemicals Supplier No. TRCD228455500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25724-58-7
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym 1,2-Benzenedicarboxylic acid, 1-decyl 2-hexyl ester, 1,2-Benzenedicarboxylic acid, decyl hexyl ester (9CI), Phthalic acid, decyl hexyl ester (8CI), Decyl hexyl phthalate, Hexyl decyl phthalate, Phthalic acid, n-decyl-n-hexyl ester, Phthalic Acid, Decylhexyl Ester
IUPAC Name 2-O-decyl 1-O-hexyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C24H38O4/c1-3-5-7-9-10-11-12-16-20-28-24(26)22-18-14-13-17-21(22)23(25)27-19-15-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3
Formula Weight 390.277
Percent Purity >95%
Chemical Name or Material Decyl Hexyl Phthalate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.