missing translation for 'onlineSavingsMsg'
Learn More

dedicator of cytokinesis 8 Antibody (OTI12D7), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72227UV

Product Code. 18181283

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

dedicator of cytokinesis 8
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
DOCK8
Recombinant protein fragment corresponding to amino acids 84-405 of human dedicator of cytokinesis 8 (NP_982272) produced in E.coli.
0.1 mL
81704.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI12D7
Western Blot, Immunohistochemistry
dedicator of cytokinesis 8, FLJ00026, FLJ00346
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.