missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dedicator of cytokinesis 8 Antibody (OTI4F3), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72226AF594
This item is not returnable.
View return policy
Description
dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| dedicator of cytokinesis 8 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DOCK8 | |
| Full length recombinant protein of human dedicator of cytokinesis 8 (NP_982272) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI4F3 | |
| Immunohistochemistry | |
| dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
| Mouse | |
| 0.1 mL | |
| 81704 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction