Learn More
DEF6 Mouse anti-Human, Clone: OTI3C4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505320
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DEF6, or IBP, is a guanine nucleotide exchange factor for RAC and CDC42 that is highly expressed in B and T cells.Specifications
| DEF6 | |
| Monoclonal | |
| Unconjugated | |
| DEF6 | |
| IBP, SLAT, SWAP70L | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 50619 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H4E7 | |
| DEF6 | |
| Full length human recombit protein of human DEF6 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.