missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEF8 Antibody (OTI4B6), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71969AF594
This item is not returnable.
View return policy
Description
DEF8 Monoclonal specifically detects DEF8 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| DEF8 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DEF8 | |
| Full length recombinant protein of human DEF8 (NP_997397) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4B6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DEF-8, differentially expressed in FDCP 8 homolog, differentially expressed in FDCP 8 homolog (mouse), FLJ20186, MGC104349 | |
| Mouse | |
| 0.1 mL | |
| 54849 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction