missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEF8 Antibody (OTI4B6), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71969MFV610
This item is not returnable.
View return policy
Description
DEF8 Monoclonal antibody specifically detects DEF8 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| DEF8 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI4B6 | |
| Flow Cytometry | |
| DEF-8, differentially expressed in FDCP 8 homolog, differentially expressed in FDCP 8 homolog (mouse), FLJ20186, MGC104349 | |
| Full length human recombinant protein of human DEF8 (NP_997397) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 54849 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction