missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEFA1, Mouse anti-Human, Clone: 1B20, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001667-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MRTLAILAAILLVALQAQAEPLQARADEVAAAPEQIAADIPEVVVSLAWDESLAPKHPGSRKSMDCYCRIPACIAGERRYGTCIYQGRLWAFCCSpecifications
| DEFA1 | |
| Monoclonal | |
| Unconjugated | |
| defensin, alpha 1 | |
| DEF1/DEFA2/HNP-1/HP-1/MGC138393/MRS | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1B20 | |
| In 1x PBS, pH 7.4 | |
| BC027917 | |
| DEFA1 | |
| DEFA1 (AAH27917, 1 a.a. ∼ 94 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1667 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction