missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DEFB132 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16658814
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16658814 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16658814 Supplier Invitrogen™ Supplier No. PA5102341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DEFB132.

Beta-defensins (also designated BDs, or hBDs in human) are small cationic peptides with broad-spectrum antimicrobial activity against a variety of enveloped viruses, fungi and bacteria. Produced in mucosal epithelia and neutrophils of several species, beta-defensins are developmentally regulated. The family of beta-defensin proteins share a common defensin-motif that is characterized by multiple cysteine residues and a highly conserved tertiary structure. Besides playing a significant role in host immune defense, many beta-defensins also are involved in sperm maturation and capacitation. beta-defensin 32, also known as beta-defensin 132, is a 95 amino acid secreted protein that most likely contains a signal peptide sequence that requires cleavage by proteolytic enzymes in order to become biologically active.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEFB132
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DEFB132
Gene Accession No. Q7Z7B7
Gene Alias BD-32; Beta-defensin 132; beta-defensin 32; DEFB132; DEFB32; DEFB-32; defensin beta 132; defensin HEL-75; Defensin, beta 132; defensin, beta 32; HEL-75; KFLL827; RP5-1103G7.6; UNQ827; UNQ827/PRO1754
Gene Symbols DEFB132
Host Species Rabbit
Immunogen A synthesized peptide derived from human DEFB132(Accession Q7Z7B7), corresponding to amino acid residues A51-S95.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 400830
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.