Learn More
Invitrogen™ DEG15 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA598622
Product Code. 16571442
  • € 464.00 / 100µL
Description
DEG15 Polyclonal Antibody for Western Blot, ELISA
DEG15 is a trypsin-like serine endopeptidase involved in the processing of glyoxysomal higher molecular weight precursor. The dimeric form carries out the specific cleavages needed to remove PTS2-containing presequences, whereas the monomeric form degrades the removed presequences and misfolded proteins (Probable). It is not required for degradation of glyoxylate cycle enzymes during seedling development.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DEG15
Polyclonal
Unconjugated
Rabbit
Protein G
RUO
Plant
Antibody
IgG
ELISA, Western Blot
2.43 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Recombinant Arabidopsis thaliana Glyoxysomal processing protease, glyoxysomal protein (572-709AA).
100 μL
Primary
-20°C or -80°C if preferred