missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DEGS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16340785
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16340785 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16340785 Supplier Invitrogen™ Supplier No. PA583830

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PEYYDHLPQH HSWVKVLWDF VFEDSLGPYA RVKRVYRLA Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 87%.

DEGS2 encodes a bifunctional enzyme that is involvedin the biosynthesis of phytosphingolipids in human skin and inother phytosphingolipid-containing tissues. This enzyme can act asa sphingolipid delta(4)-desaturase, and also as a sphingolipidC4-hydroxylase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEGS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DEGS2
Gene Accession No. Q6QHC5
Gene Alias C14orf66; Degenerative spermatocyte homolog 2; degenerative spermatocyte homolog 2, lipid desaturase; DEGS2; delta 4-desaturase, sphingolipid 2; delta(4)-desaturase, sphingolipid 2; DES2; dihydroceramide desaturase 2; FADS8; sphingolipid 4-desaturase; sphingolipid C4-hydroxylase/delta 4-desaturase; sphingolipid C4-monooxygenase; sphingolipid delta 4 desaturase/C-4 hydroxylase; sphingolipid delta(4)-desaturase 2; sphingolipid delta(4)-desaturase/C4-hydroxylase DES2; sphingolipid delta(4)-desaturase/C4-monooxygenase DES2
Gene Symbols DEGS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DEGS2. Recombinant protein control fragment (Product #RP-95684).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123099
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.