Learn More
Sartorius™ Dehydrated Media Pad Sets
Shop All Sartorius Products
Click to view available options
:
Diameter: 50mm
Specifications
Specifications
| Product Type | Dehydrated Culture Media |
| Grid Color | White |
| Diameter (Metric) | 50 mm |