missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Benidipine, TRC

Product Code. 30354037
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354037 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354037 Supplier Toronto Research Chemicals Supplier No. TRCD22946525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dehydro Benidipine
CAS 118935-44-7
Molecular Formula C28H29N3O6
Quantity 25 mg
Synonym 2,6-Dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 3-Methyl 5-[1-(Phenylmethyl)-3-piperidinyl] Ester, 2,6-Dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid Methyl 1-(Phenylmethyl)-3-piperidinyl Ester, DH 124, 3-(1-Benzylpiperidin-3-yl) 5-Methyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate
SMILES O=C(C1=C(C)N=C(C)C(C(OC)=O)=C1C2=CC([N+]([O-])=O)=CC=C2)OC3CN(CC4=CC=CC=C4)CCC3
Molecular Weight (g/mol) 503.55
Formula Weight 503.2056
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.