missing translation for 'onlineSavingsMsg'
Learn More

Dehydro Nifedipine, TRC

Product Code. 30134273
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134273 25 mg 25mg
17913095 5 mg 5mg
17962165 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134273 Supplier Toronto Research Chemicals Supplier No. TRCD22980025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67035-22-7
Molecular Formula C17 H16 N2 O6
Molecular Weight (g/mol) 344.32
Synonym 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(2-nitrophenyl)-, 3,5-dimethyl ester, 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(2-nitrophenyl)-, dimethyl ester (9CI), B 4759, BAY-b 4759, Dehydronifedipine, Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate, Nifedipine Imp. A (EP), Nitrophenylpyridine Analogue of Nifedipine, Dehydro Nifedipine
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate
SMILES COC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2ccccc2[N+](=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H16N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8H,1-4H3
Formula Weight 344.1008
Percent Purity >95%
Chemical Name or Material Dimethyl 2,6-Dimethyl-4-(2-nitrophenyl)pyridine-3,5-dicarboxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.