missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydrodolichyl Diphosphate Synthase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92521-0.1ml
This item is not returnable.
View return policy
Description
Dehydrodolichyl Diphosphate Synthase Polyclonal antibody specifically detects Dehydrodolichyl Diphosphate Synthase in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Dehydrodolichyl Diphosphate Synthase | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| cis-isoprenyltransferase, cis-prenyl transferase, CIT, CPT, Dedol-PP synthase, dehydrodolichyl diphosphate synthase, EC 2.5.1.-, FLJ13102, HDSDS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-200 of human DHDDS (NP_995583.1). IKAGPMPKHIAFIMDGNRRYAKKCQVERQEGHSQGFNKLAETLRWCLNLGILEVTVYAFSIENFKRSKSEVDGLMDLARQKFSRLMEEKEKLQKHGVCIRVLGDLHLLPLDLQELIAQAVQATKNYNKCFLNVCFAYTSRHEISNAVREMAWGVEQGLLDPSDISESLLDKCLYTNRSPHP | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79947 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction