Learn More
Thermo Scientific™ Deionization Cartridge Assembly
Use this Thermo Scientific™ Deionization Cartridge Assembly in chambers with humidity that have no DI water source.
Merk:  Thermo Scientific™ 585-036-00
Overige gegevens : Gewicht : 0.90000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15543781
  • € 331.00 / 1 stuk
onze contactgegevens 020 4877000
Toevoegen aan winkelmandje
Specificatie
Specificatie
Chambers with humidity that have no DI water source
Replacement Deionization cartridge
Customer installed
Video's