missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEK Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97614C
This item is not returnable.
View return policy
Description
DEK Polyclonal specifically detects DEK in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| DEK | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| D6S231EDEK oncogene (DNA binding), DEK oncogene, protein DEK | |
| Produced in rabbits immunized with E. coli-derived Human DEK fragment. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7913 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction