missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61058
This item is not returnable.
View return policy
Description
DEK Polyclonal specifically detects DEK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| DEK | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| D6S231EDEK oncogene (DNA binding), DEK oncogene, protein DEK | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_003463.1 | |
| DEK | |
| The immunogen recognized by this antibody maps to a region between residue 325 and 375 of human DEK oncogene using the numbering given in entry NP_003463.1 (GeneID 7913). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 7913 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction