missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEK Monoclonal Antibody (715511)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561754
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
DEK is a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases.
Specifications
| DEK | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P35659 | |
| DEK | |
| Human human embryonic kidney cell line HEK293EBNA-derived recombinant human DEK, Val282-Ser375. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 715511 | |
| Unconjugated | |
| DEK | |
| 1810019E15Rik; D13H6S231E; D6S231E; DEK; DEK oncogene (DNA binding); DEK proto-oncogene; protein DEK | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7913 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction