missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DEK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551297
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total DEK protein.
DEK is a protein with one SAP domain. This protein binds to cruciform and superhelical DNA and induces positive supercoils into closed circular DNA, and is also involved in splice site selection during mRNA processing. Chromosomal aberrations involving this region, increased expression of this gene, and the presence of antibodies against this protein are all associated with various diseases.
Specifications
| DEK | |
| Polyclonal | |
| Unconjugated | |
| DEK | |
| 1810019E15Rik; D13H6S231E; D6S231E; DEK; DEK oncogene (DNA binding); DEK proto-oncogene; protein DEK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7913 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P35659 | |
| DEK | |
| Synthetic peptide of human DEK. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction