Learn More
Delgocitinib, MedChemExpress
Product Code. 16446987
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. 16446987
Brand: Medchem Express HY109053
MedChemExpress Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively.
Specifications
Chemical Name or Material Delgocitinib
CAS 1263774-59-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H18N6O
Quantity 5 mg
Synonym JTE-052
Solubility Information DMSO : ≥ 58 mg/mL (186.89 mM)
SMILES O=C(N(C[C@@H]1C)[C@]21CN(C3=C(C=CN4)C4=NC=N3)CC2)CC#N
Molecular Weight (g/mol) 310.35
Formula Weight 310.35
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only