missing translation for 'onlineSavingsMsg'
Learn More

Delmadinone Acetate, TRC

Product Code. 30005434
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005434 10 mg 10mg
18073140 50 mg 50mg
18071950 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005434 Supplier Toronto Research Chemicals Supplier No. TRCD23066510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13698-49-2
Molecular Formula C23 H27 Cl O4
Molecular Weight (g/mol) 402.91
Synonym 17-(Acetyloxy)-6-chloropregna-1,4,6-triene-3,20-dione, Pregna-1,4,6-triene-3,20-dione, 6-chloro-17-hydroxy-, acetate (6CI,7CI,8CI), 17-(Acetyloxy)-6-chloropregna-1,4,6-triene-3,20-dione, 17-Acetoxy-6-chloropregna-1,4,6-triene-3,20-dione, 17α-Acetoxy-6-chloropregna-1,4,6-triene-3,20-dione, 6-Chloro-17-hydroxypregna-1,4,6-triene-3,20-dione acetate, Delmadinone acetate, Delminal, Estrex, RS 1301, Tardastrex, Tarden, Zenadrex, δ1-Chlormadinone acetate
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-6-chloro-10,13-dimethyl-3-oxo-9,11,12,14,15,16-hexahydro-8H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C)C(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C23H27ClO4/c1-13(25)23(28-14(2)26)10-7-18-16-12-20(24)19-11-15(27)5-8-21(19,3)17(16)6-9-22(18,23)4/h5,8,11-12,16-18H,6-7,9-10H2,1-4H3/t16-,17+,18+,21-,22+,23+/m1/s1
Formula Weight 402.1598
Chemical Name or Material Delmadinone Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.