missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ delta C2-alpha Tubulin Recombinant Rabbit Monoclonal Antibody (RM447)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544592
This item is not returnable.
View return policy
Description
This antibody reacts to cAMP (Cyclic Adenosine Monophosphate). No cross reactivity with other cyclic nucleotide or nucleoside phosphate.
Delta C2-alpha Tubulin has been identified as a biomarker for peripheral neuropathy. Delta C2-alpha tubulin is a C-terminally trimmed version of alpha-tubulin that lacks a carboxy-terminal glutamyl-tyrosine group.
Specifications
| delta C2-alpha Tubulin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Q71U36 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7846 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| RM447 | |
| Unconjugated | |
| TUBA1A | |
| TUBA1A | |
| A peptide corresponding to dC2-alpha-Tubulin. The immunogen range is a synthetic peptide within amino acids 351-451 of human alpha Tubulin which lacks a carboxy-terminal glutamyl-tyrosine group. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction