Learn More
Delta-Like Protein 1 Mouse anti-Human, BUV661, Clone: MHD1-314, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 750494
Product Code. 17231835
  • ONLINE EXCLUSIVE PRICE
    € 660.00
    € 657.57 / 50µg
    Save € 2.43
    0.37% Off
Description
Delta-Like Protein 1 Monoclonal antibody specifically detects Delta-Like Protein 1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Delta-Like Protein 1
Monoclonal
Brilliant Ultraviolet 661
DLL1; DL1; Delta1; H-Delta-1; Delta-like 1; Delta
Human DLL1 Recombinant Protein
RUO
Human
IgG1 κ
Flow Cytometry
MHD1-314
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody.