missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Delta Np73 Antibody (38C674.2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24873AF700
This item is not returnable.
View return policy
Description
Delta Np73 Monoclonal specifically detects Delta Np73 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Delta Np73 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| O15350 | |
| p73 | |
| This antibody was developed against a peptide corresponding to a portion of amino acid residues 1-50 of human dNp73. Mouse and human sequences are more than 95 % identical at these amino acid residues. In mouse, alanine at position 8 is replaced by methionine. | |
| 0.1 mL | |
| Primary | |
| This antibody is reactive to Human, Mouse and Rat. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 38C674.2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| p53-related protein, p73, tumor protein p73 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 7161 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction