missing translation for 'onlineSavingsMsg'
Learn More

Delta2-Cefcapene Pivoxil, TRC

Product Code. 30361263
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361263 50 mg 50mg
30353613 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30361263 Supplier Toronto Research Chemicals Supplier No. TRCC24256050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Delta2-Cefcapene Pivoxil
Analyte or Component Names Delta2-Cefcapene Pivoxil
Shipping Condition Room Temperature
Molecular Formula C23 H29 N5 O8 S2
Synonym (6R,7R)-3-[[(Aminocarbonyl)oxy]methyl]-7-[[(2Z)-2-(2-amino-4-thiazolyl)-1-oxo-2-pentenyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylic Acid (2,2-Dimethyl-1-oxopropoxy)methyl Ester
InChI Formula InChI=1S/C23H29N5O8S2/c1-5-6-12(13-9-38-21(24)26-13)16(29)27-14-17(30)28-15(11(7-34-22(25)33)8-37-18(14)28)19(31)35-10-36-20(32)23(2,3)4/h6,8-9,14-15,18H,5,7,10H2,1-4H3,(H2,24,26)(H2,25,33)(H,27,29)/b12-6-/t14-,15?,18-/m0/s1
Solution Type Neat
SMILES CC\C=C(/C(=O)N[C@@H]1[C@@H]2SC=C(COC(=O)N)C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)\c3csc(N)n3
IUPAC Name 2,2-dimethylpropanoyloxymethyl (6S,7S)-7-[[(Z)-2-(2-aminothiazol-4-yl)pent-2-enoyl]amino]-3-(carbamoyloxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-ene-2-carboxylate
Industry Type Forensics
Molecular Weight (g/mol) 567.64
Quantity 50 mg
Formula Weight 567.1458
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.