Learn More
MedChemExpress Deltonin
Product Code. 16481698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481698 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16481698 Supplier MedChemExpress Supplier No. HYN2283
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Deltonin, a steroidal saponin, isolated from Dioscorea zingiberensis Wright, with antitumor activity; Deltonin inhibits ERK1/2 and AKT activation.
Specifications
Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 885.04 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 55659-75-1
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 50 mg/mL (56.49 mM∣Need ultrasonic)
Target ERK∣Akt∣Endogenous Metabolite
Quantity 5 mg