missing translation for 'onlineSavingsMsg'
Learn More

Demethylpiperazinyl Sildenafil Sulfonic Acid, TRC

Product Code. 30008412
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008412 500 mg 500mg
30004699 25 mg 25mg
30010187 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008412 Supplier Toronto Research Chemicals Supplier No. TRCD231235500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1357931-55-5
Molecular Formula C17 H20 N4 O5 S
Molecular Weight (g/mol) 392.43
Synonym 4-Ethoxy-3-(1-methyl-7-oxo-3-propyl-6,7-dihydro-1Hpyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonic Acid, Sildenafil Citrate Imp. D (Pharmeuropa), 3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid
IUPAC Name 4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonic acid
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H20N4O5S/c1-4-6-12-14-15(21(3)20-12)17(22)19-16(18-14)11-9-10(27(23,24)25)7-8-13(11)26-5-2/h7-9H,4-6H2,1-3H3,(H,18,19,22)(H,23,24,25)
Formula Weight 392.1154
Percent Purity >95%
Chemical Name or Material 3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.