missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dendritic Cell Marker Antibody (AP-MAB0801), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB110-85474H
This item is not returnable.
View return policy
Description
Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Dendritic Cell Marker | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| AP-MAB0801 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DCs, Dendritic cell | |
| Mouse Dendritic cells | |
| 0.1 mL | |
| This reacts with Dendritic Cell Inhibitory Receptor 2 on mouse dendritic cells. | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction