Learn More
Dengue Virus 2 Envelope Antibody (201), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05838F
Product Code. 18652987
  • € 551.00 / 0.10mL
Description
Dengue Virus 2 Envelope Monoclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Produced in mouse immunized with recombinant Dengue Virus 2 Envelope (Accession#: AAC59274.1; Met578-Gln680).
Primary
IgG1
RUO
Virus