missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus 2 Envelope Antibody [HRP] - (Domain III), (New Guinea C/PUO-218 hybrid), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-33164H
This item is not returnable.
View return policy
Description
Dengue Virus 2 Envelope Polyclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Dengue Virus 2 Envelope | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| HRP | |
| Dengue 2 envelope;Dengue envelope;envelope protein, Envelope protein E | |
| Produced in rabbits immunized with purified, recombinant Dengue Virus 2 Envelope - (Domain III), (New Guinea C/PUO-218 hybrid) (RefSeq#: AAC59274.1; Met578-Gln680). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction