missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus 2 Envelope Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 302.00 - € 504.00
Specifications
| Antigen | Dengue Virus 2 Envelope |
|---|---|
| Dilution | Western Blot 1:2000-1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Dengue Virus 2 Envelope Polyclonal antibody specifically detects Dengue Virus 2 Envelope in Virus samples. It is validated for Western BlotSpecifications
| Dengue Virus 2 Envelope | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant Dengue Virus 2 Envelope (Accession#: AAC59274.1; Met578-Gln680). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:2000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| Dengue 2 envelope;Dengue envelope;envelope protein, Envelope protein E | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title