missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Capsid Protein Antibody (05), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06425CL1
This item is not returnable.
View return policy
Description
Dengue Virus Capsid Protein Monoclonal antibody specifically detects Dengue Virus Capsid Protein in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Dengue Virus Capsid Protein | |
| Monoclonal | |
| CoraFluor 1 | |
| polyprotein gene | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus Capsid Protein (Accession#: AHF52833.1; Met1-Arg100). | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 1494449 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| ELISA, Western Blot | |
| 05 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction