missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Capsid Protein Antibody (05), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06425DL594
This item is not returnable.
View return policy
Description
Dengue Virus Capsid Protein Monoclonal antibody specifically detects Dengue Virus Capsid Protein in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Dengue Virus Capsid Protein | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 05 | |
| Western Blot, ELISA | |
| polyprotein gene | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue virus DENV-2 (Strain New Guinea C) Capsid protein/ DENV-C (AHF52833.1; Met1-Arg100). | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 1494449 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction