missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS1 protein Antibody (12), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06437MFV610
This item is not returnable.
View return policy
Description
Dengue Virus NS1 protein Monoclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for ELISA
Specifications
| Dengue Virus NS1 protein | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352). | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| ELISA | |
| 12 | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction