missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS1 protein Antibody (12), Novus Biologicals™
Mouse Monoclonal Antibody
€ 252.00 - € 448.00
Specifications
| Antigen | Dengue Virus NS1 protein |
|---|---|
| Clone | 12 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628916
|
Novus Biologicals
NBP3-06437-100ul |
100 μg |
€ 448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623829
|
Novus Biologicals
NBP3-06437-50ul |
50 μg |
€ 252.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dengue Virus NS1 protein Monoclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for ELISASpecifications
| Dengue Virus NS1 protein | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 12 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title