Learn More
Dengue Virus NS1 protein Antibody (2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06436-50ul
 View more versions of this product
Product Code. 18655508
  • € 241.00 / 50µL
Description
Dengue Virus NS1 protein Monoclonal antibody specifically detects Dengue Virus NS1 protein in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dengue Virus NS1 protein
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus NS1 protein (Accession#: AAC59275.1; Asp1-Ala352).
50 μg
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Virus
Purified
ELISA
2
ELISA 1:1000-1:2000
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.