missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus NS4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-42897
This item is not returnable.
View return policy
Description
Specifically detects NS4B in Virus samples, and it is validated for Western Blotting
Dengue Virus NS4B Polyclonal specifically detects Dengue Virus NS4B in Viral samples. It is validated for Western Blot.
Specifications
| Dengue Virus NS4B | |
| Polyclonal | |
| Western Blot 1:500-1:3000 | |
| Rabbit | |
| 27 kDa | |
| 0.1 mL | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 1494449 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of Dengue virus Type 2 NS4B protein (Dengue virus 2 (strain 16681 PDK 53)). The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction