missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Dengue Virus NS5 Antibody (02) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30750JF549
This item is not returnable.
View return policy
Description
Dengue Virus NS5 Monoclonal antibody specifically detects Dengue Virus NS5 in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Dengue Virus NS5 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Dengue NS5, Dengue NS5 protein | |
| Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dengue Virus NS5 (New Guinea C) (RefSeq#: AAC59275.1; Gly2492-Gln2787) | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| 02 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1489713 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction