missing translation for 'onlineSavingsMsg'
Learn More

Dengue Virus NS5 Antibody - (methyltransferase domain), Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-06447-100ul

 View more versions of this product

Product Code. 18644199

  • € 507.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dengue Virus NS5 Polyclonal antibody specifically detects Dengue Virus NS5 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dengue Virus NS5
Polyclonal
Western Blot 1:20000-1:50000, ELISA 1:5000-1:10000
Dengue NS5, Dengue NS5 protein
Produced in rabbits immunized with purified, recombinant Dengue virus DENV-2 (Accession#: AAC59275.1; Gly2492-Gln2787).
100 μg
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
1489713
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato